Assay Details
Target Gene Details
Entrez Gene ID: | 166824 |
Gene Name: | Ras association domain family member 6 |
Gene Aliases: |
- |
Location: |
Chr.4:73571550-73620631 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF6 | NM_001270391.1 | 10 | 3355 | NP_001257320.1 |
| NM_001270392.1 | 10 | 3235 | NP_001257321.1 | |
| NM_177532.4 | 11 | 3457 | NP_803876.1 | |
| NM_201431.2 | 11 | 3367 | NP_958834.1 | |
| XM_011531688.1 | 12 | 3433 | XP_011529990.1 | |
| XM_011531689.2 | 10 | 3257 | XP_011529991.1 | |
| XM_011531690.2 | 10 | 5993 | XP_011529992.1 | |
| XM_011531691.1 | 9 | 3085 | XP_011529993.1 | |
| XM_017007837.1 | 8 | 5337 | XP_016863326.1 | |
| AK126346.1 | 11 | 3359 | BAC86530.1 | |
| CR749283.1 | 11 | 3457 | CAH18138.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3600997 | Chr.4:73567388 - 73580796 on Build GRCh38 | Gain |
|
| nsv1001725 | Chr.4:73488333 - 73995207 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map