Assay Details
Target Gene Details
Entrez Gene ID: | 348654 |
Gene Name: | GEN1, Holliday junction 5' flap endonuclease |
Gene Aliases: |
Gen |
Location: |
Chr.2:17753687-17785920 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GEN1 | NM_001130009.2 | 3 | 434 | NP_001123481.2 |
| NM_182625.4 | 3 | 358 | NP_872431.4 | |
| XM_005262613.4 | 3 | 444 | XP_005262670.1 | |
| XM_006712005.3 | 3 | 341 | XP_006712068.1 | |
| XM_011532820.2 | 3 | 361 | XP_011531122.1 | |
| XM_011532821.2 | 3 | 451 | XP_011531123.1 | |
| XM_011532822.2 | 3 | 331 | XP_011531124.1 | |
| AK131387.1 | 3 | 306 | BAD18538.1 | |
| BC117204.1 | 2 | 256 | ||
| BC117206.1 | 2 | 256 | ||
| BX490061.1 | ||||
| CR936783.1 | 3 | 444 | ||
| DA317583.1 | 3 | 358 | ||
| DA510949.1 | 3 | 434 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891603 | Chr.2:17752895 - 18037489 on Build GRCh38 | Gain |
|
| nsv522195 | Chr.2:17726863 - 17781183 on Build GRCh38 | Loss |
|
| nsv997502 | Chr.2:17160707 - 18147073 on Build GRCh38 | Gain |
|
| esv2759031 | Chr.2:17706783 - 17793824 on Build GRCh38 | Gain |
|
| esv3589949 | Chr.2:17755879 - 18035557 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map