Assay Details
Target Gene Details
Entrez Gene ID: | 8153 |
Gene Name: | Rho family GTPase 2 |
Gene Aliases: |
ARHN, RHO7, RhoN |
Location: |
Chr.17:43025241-43032041 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RND2 | NM_005440.4 | 5 | 3853 | NP_005431.1 |
| XM_011525316.1 | 6 | 3440 | XP_011523618.1 | |
| XM_011525317.2 | 6 | 3280 | XP_011523619.1 | |
| AK096367.1 | 1 | 2490 | ||
| BU687800.1 | 1 | 317 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2672913 | Chr.17:42964983 - 43322347 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map