Assay Details
Target Gene Details
Entrez Gene ID: | 25911 |
Gene Name: | deleted in primary ciliary dyskinesia homolog (mouse) |
Gene Aliases: |
- |
Location: |
Chr.10:101588332-101609653 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPCD | NM_015448.1 | NP_056263.1 | ||
| AK290154.1 | ||||
| AL110240.1 | CAB53691.2 | |||
| AY532267.1 | AAS46252.1 | |||
| BC001082.2 | AAH01082.2 | |||
| BC031695.1 | AAH31695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053934 | Chr.10:101409846 - 101691479 on Build GRCh38 | Gain |
|
| esv2750847 | Chr.10:101295225 - 101692888 on Build GRCh38 | Gain |
|
| esv3624382 | Chr.10:101510860 - 101643510 on Build GRCh38 | Gain |
|
| nsv527185 | Chr.10:101580299 - 101608897 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map