Assay Details
Target Gene Details
Entrez Gene ID: | 10351 |
Gene Name: | ATP binding cassette subfamily A member 8 |
Gene Aliases: |
- |
Location: |
Chr.17:68867285-68955426 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCA8 | NM_001288985.1 | 40 | 5351 | NP_001275914.1 |
| NM_001288986.1 | 39 | 5175 | NP_001275915.1 | |
| NM_007168.3 | 38 | 5070 | NP_009099.1 | |
| XM_005256938.2 | 39 | 5218 | XP_005256995.1 | |
| XM_005256940.4 | 38 | 5123 | XP_005256997.1 | |
| AB020629.1 | 38 | 5029 | ||
| AK304360.1 | ||||
| BG210966.1 | 2 | 300 | ||
| BM665457.1 | ||||
| CK824290.1 | 1 | 191 |
More Information
Set Membership: |
|

Genomic Map