Assay Details
Target Gene Details
Entrez Gene ID: | 26002 |
Gene Name: | monooxygenase DBH like 1 |
Gene Aliases: |
MOX, PRO5780, dJ248E1.1 |
Location: |
Chr.6:132296055-132401534 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOXD1 | NM_015529.3 | 10 | 1547 | NP_056344.2 |
| XM_017010714.1 | 10 | 1355 | XP_016866203.1 | |
| XM_017010715.1 | 10 | 1193 | XP_016866204.1 | |
| AK074879.1 | 10 | 1262 | BAC11263.1 | |
| AK226054.1 | 6 | 644 | ||
| AL080058.1 | 6 | 605 | CAB45692.1 | |
| AY007239.1 | 9 | 1463 | AAG09636.1 | |
| AY359094.1 | 10 | 1471 | AAQ89452.1 | |
| BC018756.1 | 10 | 1506 | AAH18756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830811 | Chr.6:132302021 - 132466400 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map