Assay Details
Target Gene Details
Entrez Gene ID: | 23431 |
Gene Name: | adaptor related protein complex 4 epsilon 1 subunit |
Gene Aliases: |
CPSQ4, SPG51, STUT1 |
Location: |
Chr.15:50908569-51005900 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4E1 | NM_001252127.1 | NP_001239056.1 | ||
| NM_007347.4 | NP_031373.2 | |||
| XM_005254264.3 | XP_005254321.1 | |||
| XM_006720447.3 | XP_006720510.1 | |||
| XM_017022042.1 | XP_016877531.1 | |||
| AB030653.1 | BAA82969.1 | |||
| AF155156.2 | AAD43326.2 | |||
| AK297290.1 | ||||
| BC126308.1 | ||||
| BC130466.1 | ||||
| CR749604.1 | CAH18399.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569403 | Chr.15:50899224 - 51012176 on Build GRCh38 | Loss |
|
| nsv569398 | Chr.15:50496928 - 51002798 on Build GRCh38 | Loss |
|
| nsv1047443 | Chr.15:50452584 - 51354826 on Build GRCh38 | Gain |
|
| nsv516168 | Chr.15:50822240 - 50925164 on Build GRCh38 | Gain |
|
| nsv1038778 | Chr.15:50904444 - 51064531 on Build GRCh38 | Loss |
|
| dgv4526n54 | Chr.15:50582767 - 51475068 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map