Assay Details
Target Gene Details
Entrez Gene ID: | 25876 |
Gene Name: | sperm flagellar 1 |
Gene Aliases: |
C20orf28, CLAMP, SPEF1A |
Location: |
Chr.20:3777504-3781466 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPEF1 | NM_015417.4 | 7 | 823 | NP_056232.2 |
| XM_005260683.4 | 7 | 782 | XP_005260740.1 | |
| AL040853.1 | 5 | 437 | ||
| AL080154.1 | 6 | 949 | CAB45745.1 | |
| BC022476.1 | 5 | 1127 | AAH22476.1 | |
| BI520334.1 | ||||
| EF560745.1 | 7 | 821 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833899 | Chr.20:3709295 - 3878350 on Build GRCh38 | Loss |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
| nsv585286 | Chr.20:3747323 - 3803946 on Build GRCh38 | Loss |
|
| nsv585285 | Chr.20:3705789 - 3791211 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map