Assay Details
Target Gene Details
Entrez Gene ID: | 27175 |
Gene Name: | tubulin gamma 2 |
Gene Aliases: |
- |
Location: |
Chr.17:42659248-42667006 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBG2 | NM_001320509.1 | NP_001307438.1 | ||
| NM_016437.2 | NP_057521.1 | |||
| XM_011524614.2 | XP_011522916.1 | |||
| XM_017024453.1 | XP_016879942.1 | |||
| XM_017024454.1 | XP_016879943.1 | |||
| XM_017024455.1 | XP_016879944.1 | |||
| XM_017024456.1 | XP_016879945.1 | |||
| XM_017024457.1 | XP_016879946.1 | |||
| AF225971.1 | AAF34188.1 | |||
| AK022324.1 | BAB14012.1 | |||
| AK225134.1 | ||||
| AK310562.1 | ||||
| BC009670.2 | AAH09670.1 | |||
| BC051890.1 | AAH51890.1 | |||
| BC108739.1 | AAI08740.1 | |||
| BE727957.1 | ||||
| BI819869.1 |
More Information
Set Membership: |
|

Genomic Map