Assay Details
Target Gene Details
Entrez Gene ID: | 5105 |
Gene Name: | phosphoenolpyruvate carboxykinase 1 |
Gene Aliases: |
PEPCK-C, PEPCK1, PEPCKC |
Location: |
Chr.20:57561081-57566461 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCK1 | NM_002591.3 | 10 | 2203 | NP_002582.3 |
| XM_011528839.1 | 8 | 1845 | XP_011527141.1 | |
| AK290802.1 | ||||
| AK300072.1 | ||||
| AW007566.1 | 1 | 490 | ||
| BC023978.1 | 10 | 2178 | AAH23978.1 | |
| BX648510.1 | 2 | 4651 | ||
| L05144.1 | 10 | 2161 | AAA60084.1 |
More Information
Set Membership: |
|

Genomic Map