Assay Details
Target Gene Details
Entrez Gene ID: | 80818 |
Gene Name: | zinc finger protein 436 |
Gene Aliases: |
ZNF, Zfp46 |
Location: |
Chr.1:23359448-23369864 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF436 | NM_001077195.1 | NP_001070663.1 | ||
| XM_011542201.1 | XP_011540503.1 | |||
| AB051497.1 |
Target Gene Details
Entrez Gene ID: | 148898 |
Gene Name: | ZNF436 antisense RNA 1 |
Gene Aliases: |
C1orf213 |
Location: |
Chr.1:23368971-23371839 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF436-AS1 | NR_033690.1 | 1 | 332 | |
| NR_033691.1 | 1 | 85 | ||
| AK074989.1 | 1 | 246 | BAC11338.1 | |
| BC028411.1 | 1 | 73 | AAH28411.1 | |
| BC054115.1 | 1 | 336 | AAH54115.1 | |
| BC068594.1 | 1 | 314 | AAH68594.1 | |
| BX098506.1 | 1 | 53 |
More Information
Set Membership: |
|

Genomic Map