Assay Details
Target Gene Details
Entrez Gene ID: | 26476 |
Gene Name: | olfactory receptor family 10 subfamily J member 1 |
Gene Aliases: |
HGMP07J, HSHGMP07J |
Location: |
Chr.1:159406348-159440967 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10J1 | NM_012351.2 | 1 | 999 | NP_036483.2 |
| XM_006711271.2 | 2 | 1011 | XP_006711334.1 | |
| XM_017001015.1 | 2 | 1304 | XP_016856504.1 | |
| BC069150.1 | AAH69150.1 | |||
| BC112290.1 | 1 | 1011 | AAI12291.1 |
More Information
Set Membership: |
|

Genomic Map