Assay Details
Target Gene Details
Entrez Gene ID: | 3671 |
Gene Name: | immunoglobulin superfamily containing leucine rich repeat |
Gene Aliases: |
HsT17563, Meflin |
Location: |
Chr.15:74173746-74176871 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISLR | NM_005545.3 | 2 | 1406 | NP_005536.1 |
| NM_201526.1 | 2 | 1222 | NP_958934.1 | |
| AB003184.1 | 2 | 1222 | BAA22848.1 | |
| AB024536.1 | 2 | 1406 | BAA85970.1 | |
| AK000817.1 | 2 | 1406 | ||
| AK074668.1 | 2 | 1814 | ||
| AK223267.1 | 2 | 1238 | BAD96987.1 | |
| AK225606.1 | 3 | 858 | ||
| AK297730.1 | 2 | 986 | ||
| AY358871.1 | 2 | 1482 | AAQ89230.1 | |
| BC022478.1 | 2 | 1211 | AAH22478.1 | |
| BC111013.1 | 2 | 1316 | AAI11014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss |
|
| nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain |
|
| nsv833054 | Chr.15:74165302 - 74332942 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map