Assay Details
Target Gene Details
Entrez Gene ID: | 114787 |
Gene Name: | G protein regulated inducer of neurite outgrowth 1 |
Gene Aliases: |
GRIN1 |
Location: |
Chr.5:176595802-176610130 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRIN1 | NM_052899.2 | 2 | 1287 | NP_443131.2 |
| AB067480.1 | 2 | 1296 | BAB67786.1 | |
| AL834165.1 | 2 | 542 | CAD38868.1 | |
| BC052950.1 | 2 | 1287 | AAH52950.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600346 | Chr.5:176597879 - 176669235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map