Assay Details
Target Gene Details
Entrez Gene ID: | 10370 |
Gene Name: | Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2 |
Gene Aliases: |
ASD8, MRG-1, MRG1, P35SRJ, VSD2 |
Location: |
Chr.6:139372255-139374650 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CITED2 | NM_001168388.2 | NP_001161860.1 | ||
| NM_001168389.2 | NP_001161861.2 | |||
| NM_006079.4 | NP_006070.2 | |||
| AF109161.1 | AAD10055.1 | |||
| BC004377.1 | AAH04377.1 | |||
| BG900380.1 | ||||
| BI763038.1 | ||||
| BM763150.1 | ||||
| BM851650.1 | ||||
| U65093.1 | AAC51114.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs199963335] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map