Assay Details
Target Gene Details
Entrez Gene ID: | 112724 |
Gene Name: | retinol dehydrogenase 13 (all-trans/9-cis) |
Gene Aliases: |
SDR7C3 |
Location: |
Chr.19:55038895-55069774 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDH13 | NM_001145971.1 | NP_001139443.1 | ||
| NM_138412.3 | NP_612421.1 | |||
| NR_027381.1 | ||||
| NR_027382.1 | ||||
| XM_005258473.1 | XP_005258530.1 | |||
| XM_011526407.1 | XP_011524709.1 | |||
| XM_011526408.2 | XP_011524710.1 | |||
| AK075392.1 | BAC11591.1 | |||
| AK122764.1 | ||||
| AK291579.1 | ||||
| AY358473.1 | AAQ88837.1 | |||
| BC009881.2 | AAH09881.1 | |||
| DB273341.1 | ||||
| DN996530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644829 | Chr.19:55030893 - 55138732 on Build GRCh38 | Gain |
|
| esv2667609 | Chr.19:54472514 - 55094226 on Build GRCh38 | Deletion |
|
| esv2763211 | Chr.19:55036566 - 55063539 on Build GRCh38 | Gain |
|
| esv2758772 | Chr.19:55026094 - 55139319 on Build GRCh38 | Gain |
|
| nsv833880 | Chr.19:54889952 - 55067343 on Build GRCh38 | Loss |
|
| esv2718872 | Chr.19:54472594 - 55094247 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114180071] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map