Assay Details
Target Gene Details
Entrez Gene ID: | 116085 |
Gene Name: | solute carrier family 22 member 12 |
Gene Aliases: |
OAT4L, RST, URAT1 |
Location: |
Chr.11:64590810-64603250 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A12 | NM_001276326.1 | 1 | 951 | NP_001263255.1 |
| NM_001276327.1 | 1 | 951 | NP_001263256.1 | |
| NM_144585.3 | 1 | 951 | NP_653186.2 | |
| NM_153378.2 | 2 | 840 | NP_700357.1 | |
| XM_006718430.3 | 1 | 557 | XP_006718493.1 | |
| XM_006718431.3 | 2 | 443 | XP_006718494.1 | |
| AB050269.1 | 1 | 951 | BAB68364.1 | |
| AB071863.1 | 1 | 351 | BAB96750.1 | |
| AK055737.1 | 2 | 422 | ||
| AK122599.1 | 1 | 505 | ||
| AK315061.1 | 1 | 499 | ||
| AY358183.1 | 1 | 484 | AAQ88550.1 | |
| BC026291.1 | 1 | 541 | ||
| BC053348.1 | 1 | 494 | AAH53348.1 | |
| DA631511.1 | 1 | 469 | ||
| DQ655912.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528506 | Chr.11:64577976 - 64933558 on Build GRCh38 | Loss |
|
| esv2422214 | Chr.11:64545019 - 64715946 on Build GRCh38 | Deletion |
|
| dgv1965n54 | Chr.11:64566642 - 64806117 on Build GRCh38 | Loss |
|
| nsv832188 | Chr.11:64547896 - 64717737 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map