Assay Details
Target Gene Details
Entrez Gene ID: | 8329 |
Gene Name: | histone cluster 1, H2ai |
Gene Aliases: |
H2A/c, H2AFC |
Location: |
Chr.6:27808199-27808667 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H2AI | NM_003509.2 | NP_003500.1 | ||
| AK311766.1 | ||||
| BC112254.1 | AAI12255.1 | |||
| BC112256.1 | AAI12257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890803 | Chr.6:27771601 - 27814760 on Build GRCh38 | Loss |
|
| nsv1017057 | Chr.6:27777998 - 27821905 on Build GRCh38 | Gain |
|
| nsv969366 | Chr.6:27807410 - 27808693 on Build GRCh38 | Duplication |
|
| nsv830618 | Chr.6:27625495 - 27821827 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map