Assay Details
Target Gene Details
Entrez Gene ID: | 25821 |
Gene Name: | mitochondrial tRNA translation optimization 1 |
Gene Aliases: |
CGI-02, COXPD10 |
Location: |
Chr.6:73461727-73501456 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576177 | Chr.6:73460675 - 73492915 on Build GRCh38 | Gain |
|
| nsv830693 | Chr.6:73339619 - 73506357 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map