Assay Details
Target Gene Details
Entrez Gene ID: | 26958 |
Gene Name: | coatomer protein complex subunit gamma 2 |
Gene Aliases: |
2-COP, gamma-2-COP |
Location: |
Chr.7:130506238-130668758 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPG2 | NM_012133.5 | 24 | 2771 | NP_036265.3 |
| AB047847.1 | 24 | 2681 | BAB17658.1 | |
| AF157833.1 | 24 | 2741 | AAF14271.1 | |
| AF207598.1 | 24 | 2737 | AAF19433.1 | |
| AF308295.1 | 5 | 634 | AAG48262.1 | |
| AI566718.1 | 1 | 373 | ||
| AK002086.1 | 16 | 2073 | BAA92078.1 | |
| AK022934.1 | 24 | 2668 | BAB14315.1 | |
| AK024414.1 | 17 | 2149 | ||
| BC017443.1 | 24 | 2664 | AAH17443.2 | |
| BX089636.1 | 1 | 385 | ||
| HY355769.1 | 1 | 371 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv473631 | Chr.7:130506625 - 130506625 on Build GRCh38 | Novel Sequence Insertion |
|
| nsv831131 | Chr.7:130468986 - 130656327 on Build GRCh38 | Loss |
|
| nsv831130 | Chr.7:130418592 - 130510992 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map