Assay Details
Target Gene Details
Entrez Gene ID: | 29902 |
Gene Name: | family with sequence similarity 216 member A |
Gene Aliases: |
C12orf24, HSU79274 |
Location: |
Chr.12:110468424-110490387 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM216A | NM_013300.2 | 7 | 1295 | NP_037432.2 |
| XM_005253875.4 | 7 | 805 | XP_005253932.1 | |
| XM_011538246.2 | 7 | 823 | XP_011536548.1 | |
| AK225731.1 | 7 | 1286 | ||
| BC020967.1 | 7 | 1111 | AAH20967.1 | |
| BF114636.1 | 1 | 235 | ||
| CR457001.1 | CAG33282.1 | |||
| U79274.1 | 7 | 1262 | AAB50215.1 |

Genomic Map