Assay Details
Target Gene Details
Entrez Gene ID: | 54996 |
Gene Name: | mitochondrial amidoxime reducing component 2 |
Gene Aliases: |
MOSC2 |
Location: |
Chr.1:220748246-220784815 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARC2 | NM_001317338.1 | 1 | 168 | NP_001304267.1 |
| NM_017898.4 | 1 | 168 | NP_060368.2 | |
| XM_017001581.1 | 1 | 149 | XP_016857070.1 | |
| AL136931.1 | 1 | 145 | CAB66865.1 | |
| BC011973.1 | 1 | 80 | AAH11973.1 | |
| BC016859.1 | AAH16859.1 | |||
| BP194343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv160470 | Chr.1:220745673 - 220749437 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map