Assay Details
Target Gene Details
Entrez Gene ID: | 2824 |
Gene Name: | glycoprotein M6B |
Gene Aliases: |
M6B |
Location: |
Chr.X:13770922-13938824 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPM6B | NM_001001994.2 | 3 | 615 | NP_001001994.1 |
| NM_001001995.2 | 4 | 933 | NP_001001995.1 | |
| NM_001001996.2 | 4 | 933 | NP_001001996.1 | |
| NM_001318729.1 | 3 | 615 | NP_001305658.1 | |
| NM_005278.4 | 3 | 813 | NP_005269.1 | |
| XM_005274489.4 | 3 | 812 | XP_005274546.1 | |
| XM_011545497.2 | 4 | 624 | XP_011543799.1 | |
| XM_017029432.1 | 4 | 623 | XP_016884921.1 | |
| XM_017029433.1 | 3 | 317 | XP_016884922.1 | |
| XM_017029434.1 | 3 | 340 | XP_016884923.1 | |
| AB209525.1 | 3 | 334 | BAD92762.1 | |
| AK095657.1 | 4 | 687 | BAC04600.1 | |
| AK294314.1 | 3 | 386 | ||
| AK299675.1 | 5 | 681 | ||
| BC008151.1 | 3 | 439 | AAH08151.1 | |
| BC047295.1 | 4 | 643 | AAH47295.1 | |
| BF525674.1 | 3 | 528 | ||
| BI599457.1 | ||||
| CD516484.1 | 3 | 615 | ||
| U45955.1 | 3 | 413 | AAB16888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528634 | Chr.X:13696631 - 14038859 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map