Assay Details
Target Gene Details
Entrez Gene ID: | 90806 |
Gene Name: | angel homolog 2 |
Gene Aliases: |
Ccr4d, KIAA0759L |
Location: |
Chr.1:212992182-213015875 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGEL2 | NM_001300753.1 | 9 | 2438 | NP_001287682.1 |
| NM_001300755.1 | 9 | 2536 | NP_001287684.1 | |
| NM_001300757.1 | 8 | 2191 | NP_001287686.1 | |
| NM_001300758.1 | 8 | 2187 | NP_001287687.1 | |
| NM_144567.4 | 9 | 2513 | NP_653168.2 | |
| NR_125333.1 | 7 | 2060 | ||
| XM_005273344.1 | 9 | 2534 | XP_005273401.1 | |
| XM_005273345.1 | 9 | 2580 | XP_005273402.1 | |
| XM_005273346.2 | 9 | 2333 | XP_005273403.1 | |
| XM_005273347.3 | 8 | 2205 | XP_005273404.1 | |
| XM_017002774.1 | 9 | 2462 | XP_016858263.1 | |
| XM_017002775.1 | 10 | 2761 | XP_016858264.1 | |
| XM_017002776.1 | 10 | 2785 | XP_016858265.1 | |
| XM_017002777.1 | 10 | 2831 | XP_016858266.1 | |
| XM_017002778.1 | 10 | 2574 | XP_016858267.1 | |
| AF510741.1 | 8 | 2142 | AAM44053.1 | |
| AK022855.1 | 1 | 504 | ||
| BC047469.1 | 8 | 2103 | AAH47469.1 |
More Information
Set Membership: |
|

Genomic Map