Assay Details
Target Gene Details
Entrez Gene ID: | 166336 |
Gene Name: | prickle planar cell polarity protein 2 |
Gene Aliases: |
EPM5 |
Location: |
Chr.3:64093850-64445476 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE2 | NM_198859.3 | 2 | 603 | NP_942559.1 |
| XM_011533432.2 | 3 | 397 | XP_011531734.1 | |
| XM_011533433.2 | 3 | 397 | XP_011531735.1 | |
| XM_011533434.2 | 3 | 222 | XP_011531736.1 | |
| XM_011533435.2 | 3 | 222 | XP_011531737.1 | |
| XM_011533436.2 | 2 | 455 | XP_011531738.1 | |
| XM_011533437.2 | 2 | 9525 | XP_011531739.1 | |
| XM_011533440.2 | 3 | 397 | XP_011531742.1 | |
| XM_017005798.1 | 2 | 327 | XP_016861287.1 | |
| AL833539.1 | 2 | 604 | ||
| BC119002.1 | 2 | 584 |
Target Gene Details
Entrez Gene ID: | 100874243 |
Gene Name: | PRICKLE2 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.3:64187544-64201561 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE2-AS3 | NR_046702.1 | 4 | 442 | |
| AK056491.1 | 4 | 442 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834717 | Chr.3:64086900 - 64265228 on Build GRCh38 | Gain |
|
| nsv3851 | Chr.3:64179135 - 64224047 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map