Assay Details
Target Gene Details
Entrez Gene ID: | 8190 |
Gene Name: | melanoma inhibitory activity |
Gene Aliases: |
CD-RAP |
Location: |
Chr.19:40775177-40777493 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIA | NM_001202553.1 | NP_001189482.1 | ||
| NM_006533.3 | NP_006524.1 | |||
| BC005910.1 | AAH05910.1 | |||
| BM715936.1 | ||||
| BM856478.1 | ||||
| BQ681978.1 | ||||
| BT007044.1 | AAP35693.1 | |||
| BX280685.1 | ||||
| CR541648.1 | CAG46449.1 | |||
| X75450.1 | CAA53203.1 |
Target Gene Details
Entrez Gene ID: | 100529262 |
Gene Name: | MIA-RAB4B readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.19:40775537-40796944 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIA-RAB4B | NR_037775.1 | |||
| AK302147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833832 | Chr.19:40754458 - 40828692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map