Assay Details
Target Gene Details
Entrez Gene ID: | 80003 |
Gene Name: | pecanex homolog 2 (Drosophila) |
Gene Aliases: |
PCNXL2 |
Location: |
Chr.1:232984109-233295735 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 34 - Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCNX2 | NM_014801.3 | NP_055616.3 | ||
| XM_011544278.2 | XP_011542580.2 | |||
| XM_017002391.1 | XP_016857880.1 | |||
| XM_017002392.1 | XP_016857881.1 | |||
| AB007895.1 | BAA23708.2 | |||
| AK055374.1 | ||||
| AK074078.1 | BAB84904.1 | |||
| AK090784.1 | ||||
| AK097229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832858 | Chr.1:232978557 - 233145726 on Build GRCh38 | Loss |
|
| esv2750829 | Chr.1:232459269 - 233243473 on Build GRCh38 | Gain |
|

Genomic Map