Assay Details
Target Gene Details
Entrez Gene ID: | 29796 |
Gene Name: | ubiquinol-cytochrome c reductase, complex III subunit X |
Gene Aliases: |
HSPC051, HSPC119, HSPC151, QCR9, UCCR7.2, UCRC |
Location: |
Chr.22:29767369-29770413 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UQCR10 | NM_001003684.1 | NP_001003684.1 | ||
| NM_013387.3 | NP_037519.2 | |||
| AB028598.1 | BAB20672.1 | |||
| AF112217.1 | AAF17205.1 | |||
| AF161468.1 | AAF29083.1 | |||
| AF161500.1 | AAF29115.1 | |||
| AF161536.1 | AAF29023.1 | |||
| AK225973.1 | ||||
| BC005402.1 | AAH05402.1 | |||
| BC015971.1 | AAH15971.1 | |||
| BI754905.1 | ||||
| CA311834.1 | ||||
| CB999839.1 | ||||
| CD674029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2724127 | Chr.22:29767558 - 29769677 on Build GRCh38 | Deletion |
|
| dgv2453n106 | Chr.22:29767547 - 29769763 on Build GRCh38 | Deletion |
|
| nsv817987 | Chr.22:29767537 - 29773447 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map