Assay Details
Target Gene Details
Entrez Gene ID: | 65095 |
Gene Name: | KRI1 homolog |
Gene Aliases: |
- |
Location: |
Chr.19:10553085-10566031 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRI1 | NM_023008.3 | 10 | 909 | NP_075384.3 |
| XM_011528190.2 | 9 | 840 | XP_011526492.1 | |
| AK023011.1 | 8 | 687 | BAB14357.1 | |
| AK093879.1 | 9 | 1021 | BAC04240.1 | |
| AK097181.1 | 2 | 144 | ||
| AK300780.1 | 10 | 893 | ||
| AK300850.1 | 10 | 914 | ||
| AL833189.1 | 9 | 1562 | ||
| BC002890.2 | 7 | 574 | AAH02890.3 | |
| BC009876.2 | 1 | 81 | AAH09876.2 | |
| BC112249.1 | 7 | 566 | AAI12250.1 | |
| BC112251.1 | 7 | 566 | AAI12252.1 | |
| BC143770.1 | 7 | 566 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833744 | Chr.19:10458964 - 10635033 on Build GRCh38 | Loss |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112414135] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map