Assay Details
Target Gene Details
Entrez Gene ID: | 205428 |
Gene Name: | chromosome 3 open reading frame 58 |
Gene Aliases: |
DIA1, GoPro49, HASF |
Location: |
Chr.3:143971798-143992368 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf58 | NM_001134470.1 | 3 | 2238 | NP_001127942.1 |
| NM_173552.3 | 3 | 3192 | NP_775823.1 | |
| XM_011512543.2 | 2 | 2777 | XP_011510845.1 | |
| XM_017005869.1 | 3 | 2072 | XP_016861358.1 | |
| AK025119.1 | 1 | 801 | ||
| AK095063.1 | 1 | 1557 | ||
| AK095161.1 | 3 | 2254 | ||
| AK095400.1 | 3 | 2098 | ||
| AL832839.1 | 3 | 2254 | ||
| BC037293.2 | 3 | 2919 | AAH37293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893777 | Chr.3:143882155 - 144016702 on Build GRCh38 | Gain |
|
| nsv591922 | Chr.3:143926400 - 144106597 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map