Assay Details
Target Gene Details
Entrez Gene ID: | 54935 |
Gene Name: | dual specificity phosphatase 23 |
Gene Aliases: |
DUSP25, LDP-3, MOSP, VHZ |
Location: |
Chr.1:159780946-159782546 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP23 | NM_001319658.1 | 2 | 520 | NP_001306587.1 |
| NM_001319659.1 | 3 | 479 | NP_001306588.1 | |
| NM_017823.4 | 3 | 472 | NP_060293.2 | |
| AB164404.1 | 2 | 505 | BAD12141.1 | |
| AF086553.1 | 2 | 320 | ||
| AK000449.1 | 3 | 456 | BAA91172.1 | |
| BC001140.1 | 3 | 442 | AAH01140.1 | |
| BG823348.1 | 2 | 502 | ||
| BQ679163.1 | 3 | 491 | ||
| DA428735.1 | 2 | 523 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map