Assay Details
Target Gene Details
Entrez Gene ID: | 23439 |
Gene Name: | ATPase Na+/K+ transporting family member beta 4 |
Gene Aliases: |
- |
Location: |
Chr.X:120362085-120383249 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP1B4 | NM_001142447.2 | 8 | 1085 | NP_001135919.1 |
| NM_012069.4 | 8 | 1073 | NP_036201.1 | |
| AF158383.2 | 8 | 1058 | AAD49692.1 | |
| AF158384.2 | 8 | 1046 | AAD49693.1 | |
| BC117227.1 | 8 | 1028 | ||
| BC143404.1 | 8 | 1028 | ||
| BC143406.1 | 8 | 923 | ||
| BC143407.1 | 7 | 899 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758889 | Chr.X:120267570 - 120449094 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map