Assay Details
Target Gene Details
Entrez Gene ID: | 81839 |
Gene Name: | VANGL planar cell polarity protein 1 |
Gene Aliases: |
KITENIN, LPP2, STB2, STBM2 |
Location: |
Chr.1:115641953-115698224 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VANGL1 | NM_001172411.1 | 4 | 522 | NP_001165882.1 |
| NM_001172412.1 | 4 | 472 | NP_001165883.1 | |
| NM_138959.2 | 4 | 528 | NP_620409.1 | |
| AB057596.1 | 4 | 472 | BAB86334.1 | |
| AB075805.1 | 3 | 269 | BAB86362.1 | |
| AF481859.1 | 4 | 484 | AAO61751.1 | |
| BC032773.1 | 2 | 196 | AAH32773.1 | |
| BC065272.1 | 4 | 505 | AAH65272.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831115 | Chr.1:115661250 - 115851698 on Build GRCh38 | Gain |
|
| nsv2509 | Chr.1:115657644 - 115693403 on Build GRCh38 | Deletion |
|
| esv27976 | Chr.1:115592696 - 116135006 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map