Assay Details
Target Gene Details
Entrez Gene ID: | 8887 |
Gene Name: | Tax1 binding protein 1 |
Gene Aliases: |
CALCOCO3, T6BP, TXBP151 |
Location: |
Chr.7:27739373-27829767 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAX1BP1 | NM_001079864.2 | 2 | 227 | NP_001073333.1 |
| NM_001206901.1 | 2 | 230 | NP_001193830.1 | |
| NM_001206902.1 | 2 | 227 | NP_001193831.1 | |
| NM_006024.6 | 2 | 227 | NP_006015.4 | |
| XM_005249900.2 | 2 | 227 | XP_005249957.1 | |
| XM_017012754.1 | 2 | 227 | XP_016868243.1 | |
| AF090891.1 | 2 | 145 | AAF24016.1 | |
| AF268075.1 | 1 | 45 | AAG03025.1 | |
| AK000098.1 | 2 | 140 | ||
| AK296720.1 | 2 | 176 | ||
| AK299729.1 | 2 | 230 | ||
| AK301064.1 | 2 | 166 | ||
| AK314292.1 | 2 | 197 | ||
| AL136586.1 | 2 | 133 | CAB66521.1 | |
| BC050358.2 | 2 | 112 | AAH50358.1 | |
| BP280714.1 | 2 | 203 | ||
| CR457056.1 | 1 | 45 | CAG33337.1 | |
| CR533556.1 | 1 | 45 | CAG38587.1 | |
| DB455556.1 | 2 | 226 | ||
| DC424592.1 | 2 | 224 | ||
| U33821.2 | 2 | 128 | AAA75595.2 |
More Information
Set Membership: |
|

Genomic Map