Assay Details
Target Gene Details
Entrez Gene ID: | 26985 |
Gene Name: | adaptor related protein complex 3 mu 1 subunit |
Gene Aliases: |
- |
Location: |
Chr.10:74120257-74151085 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3M1 | NM_001320263.1 | 11 | 4813 | NP_001307192.1 |
| NM_001320264.1 | 9 | 4652 | NP_001307193.1 | |
| NM_001320265.1 | 9 | 4364 | NP_001307194.1 | |
| NM_012095.5 | 9 | 4526 | NP_036227.1 | |
| NM_207012.3 | 10 | 4652 | NP_996895.1 | |
| NR_135191.1 | 9 | 4548 | ||
| AK001565.1 | 1 | 2006 | ||
| AV722897.1 | 1 | 453 | ||
| BU679701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551527 | Chr.10:73949148 - 74140704 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map