Assay Details
Target Gene Details
Entrez Gene ID: | 11335 |
Gene Name: | chromobox 3 |
Gene Aliases: |
HECH, HP1-GAMMA, HP1Hs-gamma |
Location: |
Chr.7:26201211-26213607 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBX3 | NM_007276.4 | NP_009207.2 | ||
| NM_016587.3 | NP_057671.2 | |||
| XM_005249611.3 | 1 | 85 | XP_005249668.1 | |
| AF136630.1 | AAF62370.1 | |||
| AL542267.3 | ||||
| BC000954.1 | AAH00954.1 | |||
| BX647444.1 | ||||
| BX648807.1 | ||||
| CD514780.1 | ||||
| CV575477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1140754 | Chr.7:26201826 - 26206366 on Build GRCh38 | Deletion |
|
| nsv606460 | Chr.7:26200084 - 26203488 on Build GRCh38 | Loss |
|
| esv2734157 | Chr.7:26201844 - 26208402 on Build GRCh38 | Deletion |
|
| nsv606459 | Chr.7:26197313 - 26203488 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map