Assay Details
Target Gene Details
Entrez Gene ID: | 202 |
Gene Name: | absent in melanoma 1 |
Gene Aliases: |
CRYBG1, ST4 |
Location: |
Chr.6:106360808-106570449 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIM1 | NM_001624.3 | NP_001615.2 | ||
| XM_005266839.3 | XP_005266896.1 | |||
| AA574200.1 | ||||
| AI765826.1 | ||||
| AK025693.1 | ||||
| U83115.1 | AAB53791.1 |
Target Gene Details
Entrez Gene ID: | 84816 |
Gene Name: | reticulon 4 interacting protein 1 |
Gene Aliases: |
NIMP, OPA10 |
Location: |
Chr.6:106559237-106630921 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTN4IP1 | XM_017011375.1 | 10 | 2250 | XP_016866864.1 |
More Information
Set Membership: |
|

Genomic Map