Assay Details
Target Gene Details
Entrez Gene ID: | 64816 |
Gene Name: | cytochrome P450 family 3 subfamily A member 43 |
Gene Aliases: |
- |
Location: |
Chr.7:99828013-99867138 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP3A43 | XM_017012542.1 | 11 | 1759 | XP_016868031.1 |
| XM_017012543.1 | 10 | 1917 | XP_016868032.1 | |
| XM_017012544.1 | 9 | 1645 | XP_016868033.1 | |
| XM_017012545.1 | 9 | 1625 | XP_016868034.1 |
More Information
Set Membership: |
|

Genomic Map