Assay Details
Target Gene Details
Entrez Gene ID: | 79841 |
Gene Name: | ATP/GTP binding protein like 2 |
Gene Aliases: |
CCP2 |
Location: |
Chr.11:47659168-47715397 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL2 | NM_024783.3 | 10 | 1319 | NP_079059.2 |
| XM_005253138.4 | 8 | 1650 | XP_005253195.1 | |
| XM_005253139.4 | 8 | 1650 | XP_005253196.1 | |
| XM_005253140.4 | 8 | 1650 | XP_005253197.1 | |
| XM_005253141.4 | 8 | 1650 | XP_005253198.1 | |
| XM_006718327.3 | 7 | 1596 | XP_006718390.1 | |
| XM_006718328.3 | 7 | 1596 | XP_006718391.1 | |
| XM_011520372.2 | 9 | 1559 | XP_011518674.1 | |
| XM_011520373.2 | 8 | 1650 | XP_011518675.1 | |
| XM_011520374.2 | 7 | 1542 | XP_011518676.1 | |
| XM_011520375.2 | 7 | 1515 | XP_011518677.1 | |
| XM_011520376.2 | 9 | 1855 | XP_011518678.1 | |
| XM_011520377.2 | 6 | 770 | XP_011518679.1 | |
| XM_011520378.2 | 9 | 1120 | XP_011518680.1 | |
| XM_011520379.2 | 8 | 915 | XP_011518681.1 | |
| XM_011520381.2 | 8 | 1650 | XP_011518683.1 | |
| XM_017018331.1 | 8 | 1650 | XP_016873820.1 | |
| XM_017018332.1 | 6 | 819 | XP_016873821.1 | |
| XM_017018333.1 | 8 | 1650 | XP_016873822.1 | |
| AK027251.1 | BAB15707.1 | |||
| AK056687.1 | 7 | 1550 | ||
| AK223171.1 | BAD96891.1 | |||
| AK223176.1 | BAD96896.1 | |||
| AK303066.1 | 9 | 1158 | ||
| AK303875.1 | 8 | 1144 | ||
| BC036234.1 | 10 | 1303 | AAH36234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469858 | Chr.11:47625963 - 47815620 on Build GRCh38 | Gain |
|
| dgv1111n100 | Chr.11:47653918 - 48644472 on Build GRCh38 | Gain |
|
| nsv832143 | Chr.11:47679256 - 47861988 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map