Assay Details
Target Gene Details
Entrez Gene ID: | 388969 |
Gene Name: | chromosome 2 open reading frame 68 |
Gene Aliases: |
HCRCN81 |
Location: |
Chr.2:85605253-85612073 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2orf68 | NM_001013649.3 | 4 | 1644 | NP_001013671.2 |
| AK092972.1 | 1 | 753 | ||
| BC111483.1 | 4 | 1617 | AAI11484.1 |
Target Gene Details
Entrez Gene ID: | 10713 |
Gene Name: | ubiquitin specific peptidase 39 |
Gene Aliases: |
65K, CGI-21, HSPC332, SAD1, SNRNP65 |
Location: |
Chr.2:85602842-85649284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP39 | NR_046347.1 | |||
| XM_006711922.1 | XP_006711985.1 | |||
| AK056334.1 | ||||
| DA548921.1 |
More Information
Set Membership: |
|

Genomic Map