Assay Details
Target Gene Details
Entrez Gene ID: | 10335 |
Gene Name: | murine retrovirus integration site 1 homolog |
Gene Aliases: |
IRAG, JAW1L |
Location: |
Chr.11:10573091-10693988 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRVI1 | NM_001098579.2 | 20 | 3051 | NP_001092049.2 |
| NM_001100163.2 | 21 | 3329 | NP_001093633.1 | |
| NM_001100167.2 | 21 | 3046 | NP_001093637.1 | |
| NM_001206880.1 | 20 | 2764 | NP_001193809.1 | |
| NM_001206881.1 | 19 | 3171 | NP_001193810.1 | |
| NM_130385.3 | 21 | 3382 | NP_569056.4 | |
| AK127209.1 | 21 | 3024 | ||
| AK304702.1 | 20 | 2398 | ||
| AL044056.1 | 1 | 332 | ||
| DA185715.1 | 1 | 206 |
Target Gene Details
Entrez Gene ID: | 100129827 |
Gene Name: | MRVI1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:10541236-10599932 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRVI1-AS1 | NR_034093.2 | |||
| NR_034094.2 | ||||
| NR_046374.1 | ||||
| NR_046375.1 | ||||
| AI075905.1 | ||||
| BC028123.1 | AAH28123.1 | |||
| BI520309.1 | ||||
| BU569149.1 | ||||
| BU570065.1 | ||||
| DB071465.1 | ||||
| DB454301.1 |
More Information
Set Membership: |
|

Genomic Map