Assay Details
Target Gene Details
Entrez Gene ID: | 79339 |
Gene Name: | olfactory receptor family 51 subfamily B member 4 |
Gene Aliases: |
HOR5'Beta1 |
Location: |
Chr.11:5301014-5301946 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51B4 | NM_033179.2 | 1 | 286 | NP_149419.2 |
| BC069094.1 | 1 | 286 | AAH69094.1 | |
| BC100919.1 | 1 | 285 | ||
| BC148186.1 | 1 | 286 | ||
| BC153815.1 | 1 | 286 |
More Information
Set Membership: |
|

Genomic Map