Assay Details
Target Gene Details
Entrez Gene ID: | 9334 |
Gene Name: | beta-1,4-galactosyltransferase 5 |
Gene Aliases: |
B4Gal-T5, BETA4-GALT-IV, beta4Gal-T5, beta4GalT-V, gt-V |
Location: |
Chr.20:49632945-49713884 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT5 | NM_004776.3 | 9 | 3977 | NP_004767.1 |
| AF289556.1 | 1 | 1795 | AAL55740.1 | |
| AK054803.1 | 1 | 1086 | ||
| AK056333.1 | 1 | 1450 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3646018 | Chr.20:49621782 - 49646113 on Build GRCh38 | Gain |
|
| nsv1065852 | Chr.20:49502654 - 49643666 on Build GRCh38 | Loss |
|
| esv3646017 | Chr.20:49610386 - 49857106 on Build GRCh38 | Gain |
|
| nsv819562 | Chr.20:49633310 - 49633696 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map