Assay Details
Target Gene Details
Entrez Gene ID: | 1999 |
Gene Name: | E74 like ETS transcription factor 3 |
Gene Aliases: |
EPR-1, ERT, ESE-1, ESX |
Location: |
Chr.1:202010519-202017188 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELF3 | NM_001114309.1 | 9 | 1714 | NP_001107781.1 |
| NM_004433.4 | 9 | 1600 | NP_004424.3 | |
| AF016295.1 | 9 | 1561 | AAB96586.1 | |
| AF017307.1 | 9 | 1649 | AAB67238.1 | |
| AF517841.1 | 9 | 1582 | AAM70481.1 | |
| AK225446.1 | ||||
| AK310814.1 | ||||
| BC003569.1 | 9 | 1568 | AAH03569.1 | |
| BX537368.1 | 8 | 4637 | CAD97611.1 | |
| U66894.1 | 9 | 1542 | AAB58075.1 | |
| U73843.1 | 9 | 1566 | AAB65823.1 | |
| U73844.1 | 10 | 1497 | AAB65824.1 | |
| U97156.1 | 9 | 1571 | AAC51884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516812 | Chr.1:202008699 - 202017290 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141014502] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map