Assay Details
Target Gene Details
Entrez Gene ID: | 382 |
Gene Name: | ADP ribosylation factor 6 |
Gene Aliases: |
- |
Location: |
Chr.14:49893018-49897054 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARF6 | NM_001663.3 | NP_001654.1 | ||
| AK313790.1 | ||||
| AL121163.1 | ||||
| BC008918.2 | AAH08918.1 | |||
| DA217686.1 | ||||
| M57763.1 | AAA90928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map