Assay Details
Target Gene Details
Entrez Gene ID: | 4943 |
Gene Name: | TBC1 domain family member 25 |
Gene Aliases: |
MG81, OATL1 |
Location: |
Chr.X:48539687-48562609 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D25 | NM_002536.2 | NP_002527.1 | ||
| AB449912.1 | ||||
| AK293521.1 | ||||
| AK293875.1 | ||||
| AK294776.1 | ||||
| AK298884.1 | ||||
| AK316199.1 | ||||
| BC101817.1 | AAI01818.1 | |||
| BC101819.1 | AAI01820.1 | |||
| BC125088.1 | ||||
| DA087200.1 | ||||
| L08240.1 | AAA59593.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map