Assay Details
Target Gene Details
Entrez Gene ID: | 84465 |
Gene Name: | multiple EGF like domains 11 |
Gene Aliases: |
- |
Location: |
Chr.15:65895296-66253737 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEGF11 | NM_032445.2 | NP_115821.2 | ||
| XM_017022670.1 | XP_016878159.1 | |||
| XM_017022671.1 | XP_016878160.1 | |||
| XM_017022672.1 | XP_016878161.1 | |||
| XM_017022673.1 | XP_016878162.1 | |||
| XM_017022674.1 | XP_016878163.1 | |||
| XM_017022675.1 | XP_016878164.1 | |||
| AB058677.1 | ||||
| AB300051.1 | ||||
| AL834326.1 | CAD38994.1 | |||
| AY358226.1 | AAQ88593.1 | |||
| BC117419.1 | ||||
| BC126313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833044 | Chr.15:65766302 - 65969046 on Build GRCh38 | Loss |
|
| nsv471684 | Chr.15:65890789 - 66030069 on Build GRCh38 | Gain |
|
| esv2760035 | Chr.15:65888447 - 66058047 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map