Assay Details
Target Gene Details
Entrez Gene ID: | 4091 |
Gene Name: | SMAD family member 6 |
Gene Aliases: |
AOVD2, HsT17432, MADH6, MADH7 |
Location: |
Chr.15:66702110-66782000 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD6 | NM_005585.4 | 1 | 508 | NP_005576.3 |
| NR_027654.1 | 1 | 508 | ||
| AF035528.1 | 1 | 525 | AAB94137.1 | |
| BU154759.1 | 1 | 636 | ||
| CA406301.1 | ||||
| CB055216.1 | 1 | 579 | ||
| CN314455.1 | 1 | 291 |
More Information
Set Membership: |
|

Genomic Map