Assay Details
Target Gene Details
Entrez Gene ID: | 134 |
Gene Name: | adenosine A1 receptor |
Gene Aliases: |
RDC7 |
Location: |
Chr.1:203127705-203167405 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADORA1 | NM_000674.2 | NP_000665.1 | ||
| NM_001048230.1 | NP_001041695.1 | |||
| XM_005244901.1 | XP_005244958.1 | |||
| XM_017000342.1 | XP_016855831.1 | |||
| AK095060.1 | ||||
| AK127752.1 | ||||
| AK289928.1 | ||||
| AK293647.1 | ||||
| AK295584.1 | ||||
| AK308733.1 | ||||
| AK316106.1 | ||||
| BC026340.1 | AAH26340.1 | |||
| DA343445.1 | ||||
| DB171678.1 | ||||
| KC884744.1 | ||||
| L22214.1 | AAA17544.1 | |||
| S45235.1 | AAB23388.1 | |||
| X68485.1 | CAA48503.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832370 | Chr.1:202997191 - 203196105 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map