Assay Details
Target Gene Details
Entrez Gene ID: | 56914 |
Gene Name: | otoraplin |
Gene Aliases: |
FDP, MIAL1 |
Location: |
Chr.20:16748353-16752164 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTOR | NM_020157.3 | 4 | 637 | NP_064542.1 |
| AF233261.1 | 5 | 579 | AAF82078.1 | |
| AF243505.1 | 4 | 595 | AAG42356.1 | |
| AJ242552.1 | 4 | 588 | CAC27443.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv90n6 | Chr.20:16750430 - 16759338 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114257218,rs200826310] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map